Benzethoniumchlorid 0.004 mol/l, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Hyamine® 1622, N-Benzyl-N,N-dimethyl-2-(2-(4-(2,4,4-trimethylpentan-2-yl)phenoxy)ethoxy)ethanaminium chloride
51126-1L-F
51126-5L-F
SIAL51126-1L-FEA
50.9
EUR
SIAL51126-1L-F
SIAL51126-5L-F
Benzethoniumchlorid 0.004 mol/l, Sigma-Aldrich®
Benzethoniumchlorid
Formel:
C₂₇H₄₂ClNO₂ |
MDL Number:
MFCD00011742 CAS-Nummer: 121-54-0 EINECS: 204-479-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...