m-Phenylendiamin Dihydrochlorid
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Benzene-1,3-diamine dihydrochloride, 1,3-Diaminobenzene dihydrochloride, 1,3-Phenylenediamine dihydrochloride, 1,3-Phenylenediammonium dichloride
SB01424.1
SB01424.10
SB01424.50
MAYBSB01424.1EA
0
EUR
MAYBSB01424.1
MAYBSB01424.10
MAYBSB01424.50
m-Phenylendiamin Dihydrochlorid
m-Phenylendiamin Dihydrochlorid
Formel:
C₆H₁₀Cl₂N₂ Schmelzpunkt: 277…278 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00012975 CAS-Nummer: 541-69-5 EINECS: 208-790-0 UN-Nummer: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...