1,3-Dichlor-2-fluor-5-nitrobenzol 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3,5-Dichloro-4-fluoro-1-nitrobenzene

H61321.06 H61321.14
H61321.06EA 56.8 EUR
H61321.06 H61321.14
1,3-Dichlor-2-fluor-5-nitrobenzol 95%
1,3-Dichloro-2-fluoro-5-nitrobenzene
Formel: C₆H₂Cl₂FNO₂
Lagerungstemperatur: Gefrierschrank
MDL Number: MFCD00129963
CAS-Nummer: 3107-19-5
EINECS: 221-466-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR