(S)-3-(Boc-amino)-3-phenylpropionsäure 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
(S)-Boc-beta3-phenylalanine, Boc-D-β-Phe-OH, (S)-3-((tert-Butoxycarbonyl)amino)-3-phenylpropanoic acid
H52072.03
H52072.MD
H52072.06
H52072.03EA
260
EUR
H52072.03
H52072.MD
H52072.06
(S)-3-(Boc-amino)-3-phenylpropionsäure 95%
(S)-3-(Boc-amino)-3-phenylpropionsäure
Purity: 95%
CAS No.: 103365-47-5
Molecular Formula: C14H19NO4
Formula Weight: 265.31
Beilstein Reference: 4695945
MDL No.: MFCD01860892
Formel:
C₁₄H₁₉NO₄ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01860892 CAS-Nummer: 103365-47-5 |