5-Methoxy-2-(trifluormethyl)pyridin ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

H32586.03 H32586.06
H32586.03EA 68.6 EUR
H32586.03 H32586.06
5-Methoxy-2-(trifluormethyl)pyridin ≥97%
5-Methoxy-2-(trifluormethyl)pyridin

Purity: 97% CAS No.: 216766-13-1 Molecular Formula: C7H6F3NO Formula Weight: 177.13 MDL No.: MFCD09864699

Formel: C₇H₆F₃NO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD09864699
CAS-Nummer: 216766-13-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR