2'-Brom-6'-fluoracetophenon 96%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-(2-Bromo-6-fluorophenyl)ethanone

H62361.03 H62361.06
H62361.03EA 251 EUR
H62361.03 H62361.06
2'-Brom-6'-fluoracetophenon 96%
2'-Brom-6'-fluoracetophenon

Purity: 96% CAS No.: 928715-37-1 Molecular Formula: C8H6BrFO Formula Weight: 217.04 Physical Form: Solid

Formel: C₈H₆BrFO
Molekulargewicht: 217.04 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD13193629
CAS-Nummer: 928715-37-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR