N-[(Benzyloxy)carbonyl]-L-tryptophan ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: (S)-2-(((Benzyloxy)carbonyl)amino)-3-(1H-indol-3-yl)propanoic acid, N(α)-Benzyloxycarbonyl-L-tryptophan, N-Cbz-L-Tryptophan, Z-Trp-OH

L07535.06 L07535.14
L07535.06EA 27.6 EUR
L07535.06 L07535.14
N-[(Benzyloxy)carbonyl]-L-tryptophan ≥98%
N(α)-Cbz-L-tryptophan

Purity: 98+% CAS No.: 7432-21-5 Molecular Formula: C19H18N2O4 Formula Weight: 338.37 Storage Temperature: 2°C to 8°C Odour: Odourless Beilstein Reference: 96743 Optical Rotation: +3.1° (c=3 in acetic acid) MDL No.: MFCD00065700

Formel: C₁₉H₁₈N₂O₄
Molekulargewicht: 338.4 g/mol
Schmelzpunkt: 121…126 °C
MDL Number: MFCD00065700
CAS-Nummer: 7432-21-5
EINECS: 231-074-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR