Kalium(iodmethyl)trifluoroborat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Potassium trifluoro(iodomethyl)borate

718947-1G 718947-5G
SIAL718947-1GEA 72.3 EUR
SIAL718947-1G SIAL718947-5G
Kalium(iodmethyl)trifluoroborat, Sigma-Aldrich®
Potassium(iodomethyl)trifluoroborate
Formel: CH₂BF₃IK
MDL Number: MFCD11112187
CAS-Nummer: 888711-47-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR