4,4'-Biphenyldiboronsäure 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L13328.03
L13328.06
L13328.03EA
217
EUR
L13328.03
L13328.06
4,4'-Biphenyldiboronsäure 95%
4,4'-Biphenyldiboronsäure
Purity: 95%
CAS No.: 4151-80-8
Molecular Formula: C12H12B2O4
Formula Weight: 241.85
Melting Point: ca 300° dec.
Beilstein Reference: 2941876
Formel:
C₁₂H₁₂B₂O₄ Schmelzpunkt: ∼ 300 °C (Zersetzung) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00151795 CAS-Nummer: 4151-80-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...