Coniferylalkohol ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
4-(3-Hydroxyprop-1-en-1-yl)-2-methoxyphenol
B24949.01
B24949.03
B24949.06
B24949.01EA
121
EUR
B24949.01
B24949.03
B24949.06
Coniferylalkohol ≥98%
Coniferylalkohol
Purity: 98%
CAS No.: 458-35-5
Molecular Formula: C10H12O3
Formula Weight: 180.20
Storage Temperature: -30°C to -10°C
Melting Point: 75-76°
Boiling Point: 163-165°/3mm
Beilstein Reference: 2048961
Merck Reference: 14,2504
MDL No.: MFCD00002922
Formel:
C₁₀H₁₂O₃ Siedepunkt: 163…165 °C (3 mmHg) Schmelzpunkt: 75…76 °C |
MDL Number:
MFCD00002922 CAS-Nummer: 458-35-5 EINECS: 207-277-9 Merck Index: 14,2504 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...