4,4'-Diaminoazobenzol 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
L03743.03
L03743.06
L03743.MD
L03743.03EA
177
EUR
L03743.03
L03743.06
L03743.MD
4,4'-Diaminoazobenzol 95%
4,4'-Diaminoazobenzol
Purity: 95%
CAS No.: 538-41-0
Molecular Formula: C12H12N4
Formula Weight: 212.26
Melting Point: ca 245° dec.
Beilstein Reference: 745553
Merck Reference: 14,2977
UN No.: 2811
MDL No.: MFCD00041892
Formel:
C₁₂H₁₂N₄ Schmelzpunkt: ∼ 245 °C (Zersetzung) Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00041892 CAS-Nummer: 538-41-0 EINECS: 208-690-7 ADR: 6.1,III Merck Index: 14,2977 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...