Methyl-2-(chlorsulfonyl)benzoat ≥94%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Methyl 2-(chlorosulphonyl)benzoate
B23211.06
B23211.14
AB131883.0005
B23211.06EA
47.6
EUR
B23211.06
B23211.14
ABCRAB131883.0005
Methyl-2-(chlorsulfonyl)benzoat ≥94%
Methyl-2-(chlorsulfonyl)benzoat
Purity: 94%
CAS No.: 26638-43-7
Molecular Formula: C8H7ClO4S
Formula Weight: 234.66
Storage Temperature: 2°C to 8°C
Melting Point: 59-64°
UN No.: 3261
MDL No.: MFCD00009797
Formel:
C₈H₇ClO₄S Schmelzpunkt: 59…64 °C |
MDL Number:
MFCD00009797 CAS-Nummer: 26638-43-7 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...