4-Brom-N,N-diphenylanilin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(4-Bromophenyl)diphenylamine, 4-Bromotriphenylamine
M-Clarity™ Qualitätsstufe = MQ100
643831-25G
643831-5G
SIAL643831-25GEA
143
EUR
SIAL643831-25G
SIAL643831-5G
4-Brom-N,N-diphenylanilin, Sigma-Aldrich®
4-Brom-N,N-diphenylanilin
Formel:
C₁₈H₁₄BrN Molekulargewicht: 324.21 g/mol |
MDL Number:
MFCD01851266 CAS-Nummer: 36809-26-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...