N(α)-Benzyloxycarbonyl-L-lysin ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
(S)-6-Amino-2-(((benzyloxy)carbonyl)amino)hexanoic acid, Z-Lys-OH
L08629.03
L08629.06
L08629.03EA
37.8
EUR
L08629.03
L08629.06
N(α)-Benzyloxycarbonyl-L-lysin ≥98%
N(α)-Benzyloxycarbonyl-L-lysin
Purity: 98+%
CAS No.: 2212-75-1
Molecular Formula: C14H20N2O4
Formula Weight: 280.32
Storage Temperature: 2°C to 8°C
Melting Point: ca 237° dec.
Beilstein Reference: 2153826
Optical Rotation: -12° (c=2 in 0.2N HCl)
MDL No.: MFCD00038204
Formel:
C₁₄H₂₀N₂O₄ Schmelzpunkt: ∼ 237 °C (dec.) |
MDL Number:
MFCD00038204 CAS-Nummer: 2212-75-1 EINECS: 218-662-6 |