(R)-3-(Boc-amino)-3-(4-bromphenyl)propionsäure 95%

Lieferant: Thermo Fisher Scientific

Synonyme: (R)-3-(4-Bromophenyl)-3-((tert-butoxycarbonyl)amino)propanoic acid

H52166.03 H52166.06
H52166.03EA 109 EUR
H52166.03 H52166.06
(R)-3-(Boc-amino)-3-(4-bromphenyl)propionsäure 95%
(R)-3-(Boc-amino)-3-(4-bromphenyl)propionsäure

Purity: 95% CAS No.: 261380-20-5 Molecular Formula: C14H18BrNO4 Formula Weight: 344.20 Melting Point: 143-145° MDL No.: MFCD01320860

Formel: C₁₄H₁₈BrNO₄
Molekulargewicht: 344.20 g/mol
Schmelzpunkt: 143…145 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01320860
CAS-Nummer: 261380-20-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR