N-Boc-L-valin ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: Boc-Val-OH, BOC-L-Valine

A16007.06 A16007.14
A16007.06EA 36.6 EUR
A16007.06 A16007.14
N-Boc-L-valin ≥98%
N-Boc-L-valin

Purity: 98+% CAS No.: 13734-41-3 Molecular Formula: C10H19NO4 Formula Weight: 217.27 Storage Temperature: 2°C to 8°C Melting Point: 77-80° Beilstein Reference: 1711290 Optical Rotation: -7.5° (c=1 in acetic acid) MDL No.: MFCD00065605

Formel: C₁₀H₁₉NO₄
Molekulargewicht: 217.3 g/mol
Schmelzpunkt: 77…80 °C
MDL Number: MFCD00065605
CAS-Nummer: 13734-41-3
EINECS: 237-307-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR