5-Nitro-2-(1-pyrrolyl)benzonitril
Lieferant: Thermo Scientific
BTB02905.1
BTB02905.10
MAYBBTB02905.1EA
0
EUR
MAYBBTB02905.1
MAYBBTB02905.10
5-Nitro-2-(1-pyrrolyl)benzonitril
5-Nitro-2-(1-pyrrolyl)benzonitril
Formel:
C₁₁H₇N₃O₂ |
MDL Number:
MFCD00052251 CAS-Nummer: 106981-59-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...