1-Boc-isonipecotinsäureethylester ≥97%
Lieferant: Thermo Fisher Scientific
Synonyme:
Ethyl N-BOC-4-piperidinecarboxylate, Ethyl N-Boc-piperidine-4-carboxylate
B25393.06
B25393.14
B25393.06EA
65.3
EUR
B25393.06
B25393.14
1-Boc-isonipecotinsäureethylester ≥97%
1-Boc-isonipecotinsäureethylester
Purity: 97+%
CAS No.: 142851-03-4
Molecular Formula: C13H23NO4
Formula Weight: 257.33
Storage Temperature: 2°C to 8°C
Flash Point:
Formel:
C₁₃H₂₃NO₄ Molekulargewicht: 257.33 g/mol Siedepunkt: 120…135 °C (0.5 mmHg) Dichte: 1.046 Flammpunkt: >110 °C (230 °F) |
MDL Number:
MFCD01763998 CAS-Nummer: 142851-03-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...