2,2'-Bichinolin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

B35407-1G B35407-5G
SIALB35407-1GEA 62.4 EUR
SIALB35407-1G SIALB35407-5G
2,2'-Bichinolin, Sigma-Aldrich®
2,2'-Bichinolin
Formel: C₁₈H₁₂N₂
MDL Number: MFCD00006740
CAS-Nummer: 119-91-5
EINECS: 204-357-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR