4-(Morpholin-4-ylmethyl)benzonitril

Lieferant: Thermo Scientific

Synonyme: 4-(Morpholinomethyl)benzonitrile

CC29316.1 CC29316.10
MAYBCC29316.1EA 0 EUR
MAYBCC29316.1 MAYBCC29316.10
4-(Morpholin-4-ylmethyl)benzonitril
4-(Morpholin-4-ylmethyl)benzonitril
Formel: C₁₂H₁₄N₂O
MDL Number: MFCD00454271
CAS-Nummer: 37812-51-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR