1-Acetylpiperidin-4-carbonsäure

Lieferant: Thermo Scientific

CD09938.1 CD09938.10 CD09938.50
MAYBCD09938.1EA 0 EUR
MAYBCD09938.1 MAYBCD09938.10 MAYBCD09938.50
1-Acetylpiperidin-4-carbonsäure
1-Acetylpiperidin-4-carbonsäure
Formel: C₈H₁₃NO₃
Molekulargewicht: 171.19 g/mol
MDL Number: MFCD00023706
CAS-Nummer: 25503-90-6
EINECS: 247-049-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR