L-Methioninsulfoxid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
H-Met(O)-OH, (2S)-2-Amino-4-(methylsulphinyl)butanoic acid
M-Clarity™ quality level = MQ200
M1126-5G
M1126-1G
SIALM1126-5GEA
276
EUR
SIALM1126-5G
SIALM1126-1G
L-Methioninsulfoxid, Sigma-Aldrich®
L-Methioninsulfoxid
Formel:
C₅H₁₁NO₃S |
MDL Number:
MFCD00063093 CAS-Nummer: 3226-65-1 EINECS: 221-758-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...