L-Methioninsulfon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (S)-2-Amino-4-(methylsulphonyl)butanoic acid

M-Clarity™ quality level = MQ200

M0876-1G M0876-5G
SIALM0876-1GEA 82.7 EUR
SIALM0876-1G SIALM0876-5G
L-Methioninsulfon, Sigma-Aldrich®
L-Methioninsulfon
Formel: C₅H₁₁NO₄S
MDL Number: MFCD00066020
CAS-Nummer: 7314-32-1
EINECS: 230-774-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR