DL-Ethionin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-Amino-4-(ethylthio)butanoic acid
M-Clarity™ quality level = MQ200
E5139-1G
E5139-25G
E5139-5G
SIALE5139-1GEA
99.8
EUR
SIALE5139-1G
SIALE5139-25G
SIALE5139-5G
DL-Ethionin, Sigma-Aldrich®
DL-Ethionin
Formel:
C₆H₁₃NO₂S |
MDL Number:
MFCD00063102 CAS-Nummer: 67-21-0 EINECS: 200-647-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...