DL-Ethionin ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Amino-4-(ethylthio)butanoic acid

A13154.03 A13154.06 A13154.14
A13154.03EA 49 EUR
A13154.03 A13154.06 A13154.14
DL-Ethionin ≥98%
DL-Ethionin

Purity: 98% CAS No.: 67-21-0 Molecular Formula: C6H13NO2S Formula Weight: 163.24 Odour: Odourless Melting Point: ca 265° dec. Beilstein Reference: 1722529 Merck Reference: 14,3738 MDL No.: MFCD00063102

Formel: C₆H₁₃NO₂S
Schmelzpunkt: ∼ 265 °C (dec.)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00063102
CAS-Nummer: 67-21-0
EINECS: 200-647-0
Merck Index: 14,3738

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR