Pallets and sumps for drums, CW-60
Lieferant: LACONT UMWELTTECHNIK
P64-2051-A P64-1051-A P64-2053-A P64-2052-A P64-1052-A P64-1053-A
LACOP64-2051-AEA 0 EUR
LACOP64-2051-A LACOP64-1051-A LACOP64-2053-A LACOP64-2052-A LACOP64-1052-A LACOP64-1053-A
Pallets and sumps for drums, CW-60
Fasslager
These pallets and sumps made of 3 mm sheet steel are ideal for the storage of water-endangering and flammable liquids in containers and drums up to 60 L.
  • Removable galvanised grid
  • Ground clearance 100 mm for internal transport
  • Corrosion protection by primer coating and painting or by fire-galvanised sheet steel