2-Mercaptothiophen

Lieferant: Thermo Scientific

Synonyme: Thiophene-2-thiol

KM01273.1 KM01273.10 KM01273.50
MAYBKM01273.1EA 0 EUR
MAYBKM01273.1 MAYBKM01273.10 MAYBKM01273.50
2-Mercaptothiophen
2-Mercaptothiophene
Formel: C₄H₄S₂
Molekulargewicht: 116.20 g/mol
MDL Number: MFCD00051666
CAS-Nummer: 7774-74-5
EINECS: 231-881-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR