(4-Chlorphenylthio)acetonitril

Lieferant: Thermo Scientific

Synonyme: [(4-Chlorophenyl)sulphanyl]acetonitrile

KM00412.1 KM00412.10 KM00412.50
MAYBKM00412.1EA 0 EUR
MAYBKM00412.1 MAYBKM00412.10 MAYBKM00412.50
(4-Chlorphenylthio)acetonitril
(4-Chlorphenylthio)acetonitril
Formel: C₈H₆ClNS
MDL Number: MFCD00019832
CAS-Nummer: 18527-19-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR