Methyl-1,2,4-triazol-3-carboxylat ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Methyl 1H-1,2,4-triazole-3-carboxylate, Methyl 1H-1,2,4-triazole-5-carboxylate

B24750.06 B24750.14 B24750.22
B24750.06EA 46.1 EUR
B24750.06 B24750.14 B24750.22
Methyl-1,2,4-triazol-3-carboxylat ≥98%
Methyl-1,2,4-triazol-3-carboxylat

Purity: 98% CAS No.: 4928-88-5 Molecular Formula: C4H5N3O2 Formula Weight: 127.10 MDL No.: MFCD00135989

Formel: C₄H₅N₃O₂
Molekulargewicht: 127.10 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00135989
CAS-Nummer: 4928-88-5
EINECS: 480-470-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR