trans-Bis(tributylstannyl)ethylen 96%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
L20130.03
L20130.06
L20130.03EA
107
EUR
L20130.03
L20130.06
trans-Bis(tributylstannyl)ethylen 96%
trans-Bis(tributylstannyl)ethylen
Purity: 96%
CAS No.: 14275-61-7
Molecular Formula: C26H56Sn2
Formula Weight: 606.15
Storage Temperature: 2°C to 8°C
Refractive Index: 1.5020
Boiling Point: 185°/0.01mm
Beilstein Reference: 3613803
UN No.: 2788
MDL No.: MFCD01631299
Formel:
C₂₆H₅₆Sn₂ Siedepunkt: 185 °C (0,01 mmHg) Dichte: 1,140 |
MDL Number:
MFCD01631299 CAS-Nummer: 14275-61-7 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...