1-(2-Cyanphenyl)piperidin

Lieferant: Thermo Scientific

Synonyme: 2-(1-Piperidinyl)benzonitrile, 2-Piperidinobenzonitrile

BTB03852.1 BTB03852.10 BTB03852.50
MAYBBTB03852.1EA 0 EUR
MAYBBTB03852.1 MAYBBTB03852.10 MAYBBTB03852.50
1-(2-Cyanphenyl)piperidin
1-(2-Cyanphenyl)piperidin
Formel: C₁₂H₁₄N₂
MDL Number: MFCD00049221
CAS-Nummer: 72752-52-4
EINECS: 427-330-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR