4-(4-Chlorphenoxy)piperidin ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

H50944.03 H50944.MD
H50944.03EA 312 EUR
H50944.03 H50944.MD
4-(4-Chlorphenoxy)piperidin ≥98%
4-(4-Chlorphenoxy)piperidin

CAS No.: 97839-99-1 Molecular Formula: C11H14ClNO Formula Weight: 211.69 MDL No.: MFCD04113544

Formel: C₁₁H₁₄ClNO
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD04113544
CAS-Nummer: 97839-99-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR