3-Propylidenphthalid cis- und trans-Gemisch ≥96%, FG, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ300
W295205-100G-K
W295205-1KG-K
W295205-5KG-K
SIALW295205-100G-KEA
83.2
EUR
SIALW295205-100G-K
SIALW295205-1KG-K
SIALW295205-5KG-K
3-Propylidenphthalid cis- und trans-Gemisch ≥96%, FG, Sigma-Aldrich®
3-Propylidenphthalid
Formel:
C₁₁H₁₀O₂ |
MDL Number:
MFCD00051813 CAS-Nummer: 17369-59-4 EINECS: 241-402-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...