Bis(benzonitril)dichlorpalladium(II) 97+%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Bis(benzonitrile)palladium chloride, Dichlorobis(benzonitrile)palladium(II), Bis(benzonitrile)palladium(II) Dichloride

207900010 207900100
ACRO207900010EA 91.1 EUR
ACRO207900010 ACRO207900100
Bis(benzonitril)dichlorpalladium(II) 97+%
Bis(benzonitril)dichlorpalladium(II)
Formel: C₁₄H₁₀Cl₂N₂Pd
Schmelzpunkt: 120 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00013123
CAS-Nummer: 14220-64-5
EINECS: 238-085-3
UN-Nummer: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to orange to brown Powder
Infrared spectrum Conforms
Elemental Analysis ≥97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR