3-(Methylsulfanyl)-1-hexanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-(Methylthio)hexan-1-ol, 3-(Methylthio)hexanol

M-Clarity™ quality level = MQ300

W343803-100G-K W343803-1KG-K
SIALW343803-100G-KEA 218 EUR
SIALW343803-100G-K SIALW343803-1KG-K
3-(Methylsulfanyl)-1-hexanol, Sigma-Aldrich®
3-(Methylthio)-1-hexanol
Formel: C₄H₁₀OS
MDL Number: MFCD00010674
CAS-Nummer: 51755-66-9
EINECS: 257-380-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR