6-Brom-4-methylchinolin 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H33298.MD
H33298.03
H33298.MDEA
52.7
EUR
H33298.MD
H33298.03
6-Brom-4-methylchinolin 96%
6-Bromo-4-methylquinoline
Purity: 96%
CAS No.: 41037-28-9
Molecular Formula: C10H8BrN
Formula Weight: 222.08
Melting Point: 94-95°
MDL No.: MFCD09258893
Formel:
C₁₀H₈BrN Schmelzpunkt: 94…95 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD09258893 CAS-Nummer: 41037-28-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...