4-Cyan-2-ethylanilin 96%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
L10732.03
L10732.06
L10732.03EA
66.2
EUR
L10732.03
L10732.06
4-Cyan-2-ethylanilin 96%
4-Cyan-2-ethylanilin
Purity: 96%
CAS No.: 170230-87-2
Molecular Formula: C9H10N2
Formula Weight: 146.19
Melting Point: 60-66°
Beilstein Reference: 7526452
UN No.: 3439
MDL No.: MFCD00041431
Formel:
C₉H₁₀N₂ Schmelzpunkt: 60…66 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00041431 CAS-Nummer: 170230-87-2 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...