2-Amino-5-brom-3,4-dimethylpyridin 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
L17672.06
L17672.03
L17672.06EA
324
EUR
L17672.06
L17672.03
2-Amino-5-brom-3,4-dimethylpyridin 97%
2-Amino-5-bromo-3,4-dimethylpyridine
Purity: 97%
CAS No.: 374537-97-0
Molecular Formula: C7H9BrN2
Formula Weight: 201.07
Melting Point: 133-135°
UN No.: 2811
MDL No.: MFCD02093947
Formel:
C₇H₉BrN₂ Schmelzpunkt: 133…135 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD02093947 CAS-Nummer: 374537-97-0 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...