4-Cyclohexylphenylboronsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Cyclohexylbenzeneboronic acid, (4-Cyclohexylphenyl)boronic acid
L18076.03
L18076.06
L18076.MD
L18076.03EA
174
EUR
L18076.03
L18076.06
L18076.MD
4-Cyclohexylphenylboronsäure ≥98%
4-Cyclohexylphenylboronsäure
Purity: 98%
CAS No.: 374538-04-2
Molecular Formula: C12H17BO2
Formula Weight: 204.08
Melting Point: ca 220°
MDL No.: MFCD02093068
Formel:
C₁₂H₁₇BO₂ Schmelzpunkt: ∼ 220 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD02093068 CAS-Nummer: 374538-04-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...