N-(2-Bromethyl)-4-tert-butylbenzolsulfonamid

Lieferant: Thermo Scientific

BTB09536.250 BTB09536.1
MAYBBTB09536.250EA 0 EUR
MAYBBTB09536.250 MAYBBTB09536.1
N-(2-Bromethyl)-4-tert-butylbenzolsulfonamid
N-(2-Bromethyl)-4-tert-butylbenzolsulfonamid
Formel: C₁₂H₁₈BrNO₂S
CAS-Nummer: 246236-63-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR