Natriumthiophenolat, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ quality level = MQ200
418293-10G
418293-50G
SIAL418293-10GEA
95.4
EUR
SIAL418293-10G
SIAL418293-50G
Natriumthiophenolat, Sigma-Aldrich®
Natriumthiophenolat
Formel:
C₆H₅NaS |
MDL Number:
MFCD00066460 CAS-Nummer: 930-69-8 EINECS: 213-224-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...