Methyl-2-(methylthio)benzoat
Lieferant: Thermo Scientific
KM06105.1
KM06105.10
MAYBKM06105.1EA
0
EUR
MAYBKM06105.1
MAYBKM06105.10
Methyl-2-(methylthio)benzoat
Methyl-2-(methylthio)benzoat
Formel:
C₉H₁₀O₂S |
MDL Number:
MFCD00068039 CAS-Nummer: 3704-28-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...