3,4-Diaminobenzonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

653845-10G 653845-1G
SIAL653845-10GEA 246 EUR
SIAL653845-10G SIAL653845-1G
3,4-Diaminobenzonitril, Sigma-Aldrich®
3,4-Diaminobenzonitril
Formel: C₇H₇N₃
MDL Number: MFCD00723901
CAS-Nummer: 17626-40-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR