2-Chlor-3-(trifluormethyl)chinoxalin 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H50549.03
H50549.06
H50549.03EA
76.8
EUR
H50549.03
H50549.06
2-Chlor-3-(trifluormethyl)chinoxalin 96%
2-Chloro-3-(trifluoromethyl)quinoxaline
Purity: 96%
CAS No.: 254732-51-9
Molecular Formula: C9H4ClF3N2
Formula Weight: 232.59
MDL No.: MFCD00243891
Formel:
C₉H₄ClF₃N₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00243891 CAS-Nummer: 254732-51-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...