Chinoxalin-6-amin ≥97%

Lieferant: Thermo Scientific

CC11714.250 38894
MAYBCC11714.250EA 0 EUR
MAYBCC11714.250 MAYB38894
Chinoxalin-6-amin ≥97%
Chinoxalin-6-amin
Formel: C₈H₇N₃
MDL Number: MFCD00462821
CAS-Nummer: 6298-37-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR