5-(Ethylthio)-1H-tetrazol (ETT) 0.25M in Acetonitril, BioSyn™ Aktivatorreagenz, zur DNA/RNA Synthese, Burdick & Jackson™
Lieferant: Honeywell Chemicals
Gefahr
Synonyme: 5-Ethyltio-1H-Tetrazole
BR721-2.5L BR721-4X2.5L
HONCBR721-2.5LEA 0 EUR
HONCBR721-2.5L HONCBR721-4X2.5L
5-(Ethylthio)-1H-tetrazol (ETT) 0.25M in Acetonitril, BioSyn™ Aktivatorreagenz, zur DNA/RNA Synthese, Burdick & Jackson™
5-(Ethylthio)-1H-tetrazol (ETT)
Reagents are specifically processed and purified to ensure the high synthesis efficiency
  • Ideal for oligonucleotide synthesis
  • Extremely low water content
  • High purity reagents
  • Low amine specifications
Reagents are passed through 0.1 µm filter prior to packaging to minimize particulate contamination.
Formel: C₃H₆N₄S
Siedepunkt: 82 °C
Dichte: 0.790 g/cm3
Flammpunkt: 7 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00068733
CAS-Nummer: 89797-68-2
UN-Nummer: 1648
ADR: 3,II