4-Chlor-6-chinolinboronsäurepinakolester 96%

Lieferant: Thermo Fisher Scientific

H54085.MD H54085.03
H54085.MDEA 104 EUR
H54085.MD H54085.03
4-Chlor-6-chinolinboronsäurepinakolester 96%
4-Chlor-6-chinolinboronsäurepinakolester

Purity: 96% Molecular Formula: C15H17BClNO2 Formula Weight: 289.56 MDL No.: MFCD16987803

Formel: C₁₅H₁₇BClNO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD16987803
CAS-Nummer: 1201844-73-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR