3-(Methylsulfonyl)phenylboronsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-(Methanesulphonyl)phenylboronic acid, 3-(Methylsulphonyl)benzeneboronic acid, (3-(Methylsulphonyl)phenyl)boronic acid
H52916.03
H52916.06
H52916.03EA
161
EUR
H52916.03
H52916.06
3-(Methylsulfonyl)phenylboronsäure ≥98%
3-(Methylsulfonyl)phenylboronsäure
Purity: 98%
CAS No.: 373384-18-0
Molecular Formula: C7H9BO4S
Formula Weight: 200.02
Melting Point: 102-110°
MDL No.: MFCD03092935
Formel:
C₇H₉BO₄S Schmelzpunkt: 102…110 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03092935 CAS-Nummer: 373384-18-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...