Cer(III)oxalat Hydrat ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Cerium(III) oxalate hydrate
40227.14
40227.22
40227.14EA
94.1
EUR
40227.14
40227.22
Cer(III)oxalat Hydrat ≥99%
Cer(III)oxalat Hydrat
Purity: 99%
CAS No.: 15750-47-7
Molecular Formula: Ce2(C2O4)3.xH2O
Formula Weight: 544.29(anhy)
Physical Form: Crystalline powder
Sensitivity: Hygroscopic
Solubility: Insoluble in water. Soluble in hot dilute HCl and H2SO4
Merck Reference: 14,2001
UN No.: 3288
MDL No.: MFCD00150139
Formel:
C₆Ce₂O₁₂·H₂O Schmelzpunkt: Dec. Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00150139 CAS-Nummer: 15750-47-7 ADR: 6.1,III Merck Index: 14,2001 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...