N-Fmoc-L-norleucin
Lieferant: Novabiochem (Merck Biosciences)
Synonyme:
Fmoc-Nle-OH, FMOC-L-Norleucine
8.52014.0025
8.52014.0005
04-12-1028-25EA
369
EUR
04-12-1028-25
04-12-1028-5
N-Fmoc-L-norleucin
N-Fmoc-L-norleucin
Formel:
C₂₁H₂₃NO₄ Molekulargewicht: 353.4 g/mol |
MDL Number:
MFCD00037537 CAS-Nummer: 77284-32-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...